siRNA Details
| Gene Symbol: Efhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109310.1 | NP_001102780.1 | 4 | 700 | ||
| XM_006245468.2 | XP_006245530.1 | 5 | 779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efhd1| Gene Symbol: Efhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109310.1 | NP_001102780.1 | 4 | 700 | ||
| XM_006245468.2 | XP_006245530.1 | 5 | 779 | |||