siRNA Details
| Gene Symbol: Rbm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767346.1 | XP_008765568.1 | 8, 9 | 2227 | ||
| XM_576610.6 | XP_576610.2 | 8, 9 | 2368 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm44| Gene Symbol: Rbm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767346.1 | XP_008765568.1 | 8, 9 | 2227 | ||
| XM_576610.6 | XP_576610.2 | 8, 9 | 2368 | |||