siRNA Details
| Gene Symbol: RGD1562136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109086.1 | NP_001102556.1 | 3 | 909 | ||
| GenBank mRNA | FQ210387.1 | - | 3 | 840 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562136| Gene Symbol: RGD1562136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109086.1 | NP_001102556.1 | 3 | 909 | ||
| GenBank mRNA | FQ210387.1 | - | 3 | 840 | ||