siRNA Details
Gene Symbol: Vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109312.1 | NP_001102782.1 | 2 | 455 | ||
XM_008770349.1 | XP_008768571.1 | 2 | 742 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109312.1 | NP_001102782.1 | 2 | 455 | ||
XM_008770349.1 | XP_008768571.1 | 2 | 742 |