siRNA Details
| Gene Symbol: LOC501282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_576697.6 | XP_576697.3 | 4, 5 | 363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC501282| Gene Symbol: LOC501282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_576697.6 | XP_576697.3 | 4, 5 | 363 | ||