siRNA Details
| Gene Symbol: LOC501391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227951.2 | XP_006228013.1 | 4 | 515 | ||
| XM_008758800.1 | XP_008757022.1 | 4 | 900 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC501391| Gene Symbol: LOC501391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227951.2 | XP_006228013.1 | 4 | 515 | ||
| XM_008758800.1 | XP_008757022.1 | 4 | 900 | |||