siRNA Details
| Gene Symbol: Cbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768485.1 | XP_008766707.1 | 5 | 510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cbx4| Gene Symbol: Cbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768485.1 | XP_008766707.1 | 5 | 510 | ||