siRNA Details
| Gene Symbol: Zbtb33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109314.1 | NP_001102784.1 | 2 | 1520 | ||
| XM_006257481.2 | XP_006257543.1 | 2 | 1446 | |||
| Gene Symbol: LOC100910807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752135.3 | XP_003752183.1 | 1619 | |||
| XM_006257485.2 | XP_006257547.1 | 1446 | ||||

Zbtb33
Genomic Map