siRNA Details
| Gene Symbol: RGD1560927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109315.1 | NP_001102785.1 | 2 | 232 | ||
| Gene Symbol: LOC103694509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773491.1 | XP_008771713.1 | 2 | 251 | ||

RGD1560927
Genomic Map