siRNA Details
| Gene Symbol: Trappc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024965.1 | NP_001020136.1 | 3, 4 | 203 | ||
| XM_006256857.1 | XP_006256919.1 | 3, 4 | 293 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc2| Gene Symbol: Trappc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024965.1 | NP_001020136.1 | 3, 4 | 203 | ||
| XM_006256857.1 | XP_006256919.1 | 3, 4 | 293 | |||