siRNA Details
| Gene Symbol: Rai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109316.1 | NP_001102786.1 | 2 | 730 | ||
| XM_006256893.2 | XP_006256955.1 | 1 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rai2| Gene Symbol: Rai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109316.1 | NP_001102786.1 | 2 | 730 | ||
| XM_006256893.2 | XP_006256955.1 | 1 | 541 | |||