siRNA Details
| Gene Symbol: RGD1562161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127568.1 | NP_001121040.1 | 4, 5 | 1410 | ||
| XM_006256933.2 | XP_006256995.1 | 4, 5 | 1465 | |||
| XM_006256934.2 | XP_006256996.1 | 4, 5 | 1741 | |||
| XM_006256936.2 | XP_006256998.1 | 4, 5 | 1466 | |||
| XM_006256937.2 | XP_006256999.1 | 4, 5 | 1465 | |||
| XM_006256938.2 | XP_006257000.1 | 4, 5 | 1465 | |||
| XM_008773186.1 | XP_008771408.1 | 4, 5 | 1465 | |||
| GenBank mRNA | BC166438.1 | - | 4, 5 | 1410 | 7441789 | |

RGD1562161
Genomic Map