siRNA Details
Gene Symbol: Ctag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109322.1 | NP_001102792.1 | 1 | 74 | ||
XM_008773587.1 | XP_008771809.1 | 1 | 56 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109322.1 | NP_001102792.1 | 1 | 74 | ||
XM_008773587.1 | XP_008771809.1 | 1 | 56 |