siRNA Details
| Gene Symbol: Vma21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253301.2 | XP_006253363.1 | 383 | |||
| XM_008771344.1 | XP_008769566.1 | 654 | ||||
| GenBank mRNA | FQ211836.1 | - | 347 | |||
| FQ215711.1 | - | 649 | ||||
| Gene Symbol: LOC100912478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751684.3 | XP_003751732.1 | 3 | 383 | ||
| XM_006253887.2 | XP_006253949.1 | 4 | 654 | |||
| Gene Symbol: LOC103689964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253890.2 | XP_006253952.1 | 654 | |||
| XM_577054.6 | XP_577054.2 | 383 | ||||
| Gene Symbol: LOC103694079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771606.1 | XP_008769828.1 | 217 | |||

Vma21
Genomic Map