siRNA Details
| Gene Symbol: Fam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109324.1 | NP_001102794.1 | 3 | 475 | ||
| XM_006229603.2 | XP_006229665.1 | 2 | 632 | |||
| XM_006229604.2 | XP_006229666.1 | 2 | 634 | |||
| XM_006229605.2 | XP_006229667.1 | 2 | 633 | |||
| GenBank mRNA | BC169081.1 | - | 2 | 539 | 5621009 | |

Fam3a
Genomic Map