siRNA Details
| Gene Symbol: Fam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109324.1 | NP_001102794.1 | 8 | 832 | ||
| XM_006229603.2 | XP_006229665.1 | 7 | 989 | |||
| XM_006229604.2 | XP_006229666.1 | 7 | 3401 | |||
| XM_006229605.2 | XP_006229667.1 | 8 | 2714 | |||
| GenBank mRNA | BC169081.1 | - | 7 | 896 | 5621009 | |

Fam3a
Genomic Map