siRNA Details
| Gene Symbol: Dmrtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025288.1 | NP_001020459.1 | 1 | 70 | ||
| GenBank mRNA | BC097356.1 | - | 1 | 70 | 7442140 | |
| Gene Symbol: LOC100909986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752209.3 | XP_003752257.1 | 1, 2 | 315 | ||
| XM_006257731.2 | XP_006257793.1 | 1, 2 | 315 | |||

Dmrtc1a
Genomic Map