siRNA Details
| Gene Symbol: Lsm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173384.1 | NP_001166855.1 | 4 | 1009 | ||
| XM_006246151.2 | XP_006246213.1 | 3 | 1040 | |||
| GenBank mRNA | BC158836.1 | - | 4 | 1009 | 7385359 | |
| FQ223124.1 | - | 4 | 1045 | |||

Lsm11
Genomic Map