siRNA Details
| Gene Symbol: Lsm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173384.1 | NP_001166855.1 | 3 | 617 | ||
| GenBank mRNA | BC158836.1 | - | 3 | 617 | 7385359 | |
| FQ223124.1 | - | 3 | 653 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm11| Gene Symbol: Lsm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173384.1 | NP_001166855.1 | 3 | 617 | ||
| GenBank mRNA | BC158836.1 | - | 3 | 617 | 7385359 | |
| FQ223124.1 | - | 3 | 653 | |||