siRNA Details
| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577104.6 | XP_577104.3 | 6 | 1717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpv| Gene Symbol: Cenpv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577104.6 | XP_577104.3 | 6 | 1717 | ||