siRNA Details
| Gene Symbol: LOC103690169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773862.1 | XP_008772084.1 | 3 | 1779 | ||
| Gene Symbol: Slfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768151.1 | XP_008766373.1 | 5 | 2415 | ||

LOC103690169
Genomic Map