siRNA Details
| Gene Symbol: RGD1562771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248117.1 | XP_006248179.1 | 1 | 29 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562771| Gene Symbol: RGD1562771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248117.1 | XP_006248179.1 | 1 | 29 | ||