siRNA Details
Gene Symbol: Prr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248788.2 | XP_006248850.1 | 6 | 3270 | ||
XM_008769018.1 | XP_008767240.1 | 6 | 3612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248788.2 | XP_006248850.1 | 6 | 3270 | ||
XM_008769018.1 | XP_008767240.1 | 6 | 3612 |