siRNA Details
| Gene Symbol: Coro1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109327.1 | NP_001102797.1 | 3, 4 | 431 | ||
| GenBank mRNA | BC166764.1 | - | 3, 4 | 378 | 7932115 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coro1c| Gene Symbol: Coro1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109327.1 | NP_001102797.1 | 3, 4 | 431 | ||
| GenBank mRNA | BC166764.1 | - | 3, 4 | 378 | 7932115 | |