siRNA Details
Gene Symbol: Coro1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109327.1 | NP_001102797.1 | 5 | 643 | ||
GenBank mRNA | BC166764.1 | - | 5 | 590 | 7932115 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Coro1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109327.1 | NP_001102797.1 | 5 | 643 | ||
GenBank mRNA | BC166764.1 | - | 5 | 590 | 7932115 |