siRNA Details
| Gene Symbol: Slc2a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191551.1 | NP_001178480.1 | 7, 8 | 965 | ||
| XM_006251100.1 | XP_006251162.1 | 8, 9 | 998 | |||
| XM_006251101.2 | XP_006251163.1 | 8, 9 | 1104 | |||
| XM_006251102.2 | XP_006251164.1 | 7, 8 | 1136 | |||

Slc2a9
Genomic Map