siRNA Details
| Gene Symbol: Gzmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252044.2 | XP_006252106.2 | 9 | 1019 | ||
| XM_008770696.1 | XP_008768918.1 | 8 | 1002 | |||
| XM_008770697.1 | XP_008768919.1 | 9 | 1070 | |||
| XM_008770698.1 | XP_008768920.1 | 9 | 1045 | |||
| XM_008770699.1 | XP_008768921.1 | 8 | 1003 | |||
| XM_008770700.1 | XP_008768922.1 | 6 | 787 | |||

Gzmn
Genomic Map