siRNA Details
| Gene Symbol: LOC502176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025057.2 | NP_001020228.2 | 3 | 228 | ||
| GenBank mRNA | AY325157.1 | - | 3 | 228 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC502176| Gene Symbol: LOC502176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025057.2 | NP_001020228.2 | 3 | 228 | ||
| GenBank mRNA | AY325157.1 | - | 3 | 228 | ||