siRNA Details
| Gene Symbol: Vom2r80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111321.1 | NP_001104791.1 | 2445 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r80| Gene Symbol: Vom2r80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111321.1 | NP_001104791.1 | 2445 | |||
Set Membership: |
|