siRNA Details
| Gene Symbol: RGD1564719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109335.1 | NP_001102805.1 | 3 | 800 | ||
| XM_006228896.2 | XP_006228958.1 | 4 | 400 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564719| Gene Symbol: RGD1564719 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109335.1 | NP_001102805.1 | 3 | 800 | ||
| XM_006228896.2 | XP_006228958.1 | 4 | 400 | |||