siRNA Details
| Gene Symbol: Stard5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192011.1 | NP_001178940.1 | 1 | 50 | ||
| GenBank mRNA | FQ230578.1 | - | 1 | 24 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stard5| Gene Symbol: Stard5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192011.1 | NP_001178940.1 | 1 | 50 | ||
| GenBank mRNA | FQ230578.1 | - | 1 | 24 | ||