siRNA Details
| Gene Symbol: Eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109339.1 | NP_001102809.1 | 10 | 924 | ||
| XM_006232388.2 | XP_006232450.1 | 10 | 1071 | |||
| GenBank mRNA | FQ226213.1 | - | 3 | 373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2a| Gene Symbol: Eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109339.1 | NP_001102809.1 | 10 | 924 | ||
| XM_006232388.2 | XP_006232450.1 | 10 | 1071 | |||
| GenBank mRNA | FQ226213.1 | - | 3 | 373 | ||