siRNA Details
Gene Symbol: RGD1564854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109341.1 | NP_001102811.1 | 3 | 311 | ||
XM_006233991.2 | XP_006234053.1 | 3 | 379 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1564854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109341.1 | NP_001102811.1 | 3 | 311 | ||
XM_006233991.2 | XP_006234053.1 | 3 | 379 |