siRNA Details
| Gene Symbol: RGD1564854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109341.1 | NP_001102811.1 | 6 | 487 | ||
| XM_006233991.2 | XP_006234053.1 | 6 | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564854| Gene Symbol: RGD1564854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109341.1 | NP_001102811.1 | 6 | 487 | ||
| XM_006233991.2 | XP_006234053.1 | 6 | 555 | |||