siRNA Details
| Gene Symbol: Mettl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114181.1 | NP_001107653.1 | 6 | 781 | ||
| GenBank mRNA | BC158754.1 | - | 6 | 781 | 9102963 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl5| Gene Symbol: Mettl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114181.1 | NP_001107653.1 | 6 | 781 | ||
| GenBank mRNA | BC158754.1 | - | 6 | 781 | 9102963 | |