siRNA Details
| Gene Symbol: Cst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109343.1 | NP_001102813.1 | 2 | 280 | ||
| XM_006235162.2 | XP_006235224.1 | 3 | 871 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cst13| Gene Symbol: Cst13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109343.1 | NP_001102813.1 | 2 | 280 | ||
| XM_006235162.2 | XP_006235224.1 | 3 | 871 | |||