siRNA Details
| Gene Symbol: LOC502684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025060.1 | NP_001020231.1 | 7 | 784 | ||
| XM_006235246.2 | XP_006235308.1 | 7 | 825 | |||
| XM_006235247.2 | XP_006235309.1 | 7 | 823 | |||
| XM_006235248.2 | XP_006235310.1 | 7 | 924 | |||
| XM_006235249.2 | XP_006235311.1 | 8 | 802 | |||
| XM_006235251.2 | XP_006235313.1 | 7 | 824 | |||
| XM_008762376.1 | XP_008760598.1 | 7 | 903 | |||
| XM_008762377.1 | XP_008760599.1 | 8 | 862 | |||
| XM_008762378.1 | XP_008760600.1 | 7 | 804 | |||
| GenBank mRNA | BC083680.1 | - | 7 | 784 | 7135883 | |

LOC502684
Genomic Map