siRNA Details
| Gene Symbol: Dok5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109344.2 | NP_001102814.1 | 2 | 226 | ||
| XM_006235673.2 | XP_006235735.1 | 2 | 405 | |||
| GenBank mRNA | BC167027.1 | - | 2 | 226 | 7315978 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dok5| Gene Symbol: Dok5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109344.2 | NP_001102814.1 | 2 | 226 | ||
| XM_006235673.2 | XP_006235735.1 | 2 | 405 | |||
| GenBank mRNA | BC167027.1 | - | 2 | 226 | 7315978 | |