siRNA Details
| Gene Symbol: Ankrd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192012.1 | NP_001178941.1 | 5 | 751 | ||
| XM_006236119.2 | XP_006236181.1 | 5 | 1981 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd7| Gene Symbol: Ankrd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192012.1 | NP_001178941.1 | 5 | 751 | ||
| XM_006236119.2 | XP_006236181.1 | 5 | 1981 | |||