siRNA Details
Gene Symbol: Prrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749809.3 | XP_003749857.1 | 2 | 1281 | ||
XM_006237062.2 | XP_006237124.1 | 2 | 1359 | |||
XM_006237064.2 | XP_006237126.1 | 2 | 1204 | |||
XM_008763200.1 | XP_008761422.1 | 1 | 1203 |
