siRNA Details
| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109354.1 | NP_001102824.1 | 6 | 770 | ||
| XM_006237099.2 | XP_006237161.1 | 4 | 1372 | |||
| XM_008763325.1 | XP_008761547.1 | 4 | 796 | |||
| XM_008763326.1 | XP_008761548.1 | 5 | 495 | |||
| GenBank mRNA | GU357486.1 | - | 6 | 671 | ||

Clec9a
Genomic Map