siRNA Details
Gene Symbol: Rerg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237582.2 | XP_006237644.1 | 4 | 541 | ||
XM_578417.6 | XP_578417.4 | 4 | 546 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rerg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237582.2 | XP_006237644.1 | 4 | 541 | ||
XM_578417.6 | XP_578417.4 | 4 | 546 |