siRNA Details
| Gene Symbol: Rerg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237582.2 | XP_006237644.1 | 5 | 805 | ||
| XM_578417.6 | XP_578417.4 | 5 | 810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rerg| Gene Symbol: Rerg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237582.2 | XP_006237644.1 | 5 | 805 | ||
| XM_578417.6 | XP_578417.4 | 5 | 810 | |||