siRNA Details
| Gene Symbol: Angptl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025065.1 | NP_001020236.1 | 3 | 665 | ||
| XM_006238440.2 | XP_006238502.1 | 3 | 852 | |||
| GenBank mRNA | BC088192.1 | - | 3 | 665 | 7372069 | |
| FQ210243.1 | - | 3 | 675 | |||

Angptl3
Genomic Map