siRNA Details
| Gene Symbol: Sesn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109358.2 | NP_001102828.1 | 4 | 822 | ||
| GenBank mRNA | BC166423.1 | - | 4 | 822 | 7096599 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sesn2| Gene Symbol: Sesn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109358.2 | NP_001102828.1 | 4 | 822 | ||
| GenBank mRNA | BC166423.1 | - | 4 | 822 | 7096599 | |