siRNA Details
| Gene Symbol: Tmem229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109359.1 | NP_001102829.1 | 3 | 737 | ||
| XM_006240275.2 | XP_006240337.1 | 2 | 810 | |||
| XM_006240276.2 | XP_006240338.1 | 2 | 806 | |||
| XM_006240277.1 | XP_006240339.1 | 1 | 301 | |||
| XM_008764845.1 | XP_008763067.1 | 2 | 405 | |||
| GenBank mRNA | FQ211952.1 | - | 3 | 612 | ||

Tmem229b
Genomic Map