siRNA Details
| Gene Symbol: Vash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578575.6 | XP_578575.2 | 5 | 1997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vash1| Gene Symbol: Vash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_578575.6 | XP_578575.2 | 5 | 1997 | ||