siRNA Details
| Gene Symbol: Sult6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192017.1 | NP_001178946.1 | 3 | 328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult6b1| Gene Symbol: Sult6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192017.1 | NP_001178946.1 | 3 | 328 | ||