siRNA Details
| Gene Symbol: Zfp717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766011.1 | XP_008764233.1 | 12 | 1309 | ||
| XM_008766012.1 | XP_008764234.1 | 12 | 1151 | |||
| XM_008766013.1 | XP_008764235.1 | 11 | 1258 | |||
| XM_008766014.1 | XP_008764236.1 | 11 | 1100 | |||
| XM_008766015.1 | XP_008764237.1 | 12 | 1106 | |||
| XM_008766016.1 | XP_008764238.1 | 11 | 1245 | |||
| XM_008766017.1 | XP_008764239.1 | 11 | 1087 | |||

Zfp717
Genomic Map