siRNA Details
Gene Symbol: Zfp717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766011.1 | XP_008764233.1 | 11 | 1221 | ||
XM_008766012.1 | XP_008764234.1 | 11 | 1063 | |||
XM_008766013.1 | XP_008764235.1 | 10 | 1170 | |||
XM_008766014.1 | XP_008764236.1 | 10 | 1012 | |||
XM_008766015.1 | XP_008764237.1 | 11 | 1018 | |||
XM_008766016.1 | XP_008764238.1 | 10 | 1157 | |||
XM_008766017.1 | XP_008764239.1 | 10 | 999 |
